2-噻吩甲酰三氟丙酮 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 4,4,4-三氟-1-(2-噻吩基)-1,3-丁二酮

138610250 138611000
ACRO138610250EA 0 CNY
ACRO138610250 ACRO138611000
2-噻吩甲酰三氟丙酮 99%
2-噻吩甲酰三氟丙酮
分子式: C₈H₅F₃O₂S
沸点: 96…98 °C (8 mmHg)
溶点: 40…44 °C
闪点: 111 °C
存储条件: Ambient
MDL编号: MFCD00005445
CAS号: 326-91-0
EINECS: 206-316-7

现在订购

产品技术规格

Appearance Light yellow to beige to brown Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 40 to 44 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR