N,N-二乙基-对苯二胺 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
4-氨基-N,N-二乙苯胺
407390050
407390250
ACRO407390050EA
0
CNY
ACRO407390050
ACRO407390250
N,N-二乙基-对苯二胺 98%
N,N-二乙基-对苯二胺
分子式:
C₁₀H₁₆N₂ 分子量: 164.25 g/mol 沸点: 260 溶点: 23 密度: 0.988 g/cm³ 闪点: 139 存储条件: Ambient |
MDL编号:
MFCD00007861 CAS号: 93-05-0 EINECS: 202-214-1 UN号: 2923 ADR: 8 III |
产品技术规格
Appearance | Clear yellow-brown to dark brown to black Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...