酚红钠盐 ACS
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
酚磺酞
417250100
417250050
ACRO417250100EA
0
CNY
ACRO417250100
ACRO417250050
酚红钠盐 ACS
酚红钠盐
分子式:
C₁₉H₁₃NaO₅S 分子量: 376.35 g/mol 溶点: 285 °C (dec.) 存储条件: Ambient |
MDL编号:
MFCD00066901 CAS号: 34487-61-1 EINECS: 252-057-8 Merck Index: 12,07397 |
产品技术规格
Appearance | Dark red to red-brown Powder |
Infrared spectrum | Conforms |
Visual transition interval | From pH 6.8 (yellow) to pH 8.2 (red) |
Clarity of solution | Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...