若丹明 B 98+%, 纯净

供应商: Thermo Fisher Scientific
Danger

296570100 296570250
ACRO296570100EA 0 CNY
ACRO296570100 ACRO296570250
若丹明 B 98+%, 纯净
若丹明 B
分子式: C₂₈H₃₁ClN₂O₃
溶点: 210…211 °C (dec.)
密度: 1,31 g/cm³ (20 °C)
存储条件: Ambient
MDL编号: MFCD00011931
CAS号: 81-88-9
EINECS: 201-383-9
Merck Index: 13,08266

现在订购

产品技术规格

Appearance Dark green Powder or crystals
Infrared spectrum Conforms
HPLC ≥98.0 %
Moisture ≤2.0 % (105°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR