2-噻吩甲腈 97%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2-氰基噻吩, 噻吩-2-甲腈

298050100 298050500
ACRO298050100EA 0 CNY
ACRO298050100 ACRO298050500
2-噻吩甲腈 97%
2-噻吩甲腈
分子式: C₅H₃NS
分子量: 109.15 g/mol
沸点: 192 °C (760 mmHg)
密度: 1.17 g/cm³
闪点: 53 °C
存储条件: Ambient
MDL编号: MFCD00005416
CAS号: 1003-31-2
EINECS: 213-706-0
UN号: 1992
ADR: 3 III

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5620 to 1.5640 (20°C, 589 nm)
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR