CIP 100® Detergent System
供应商: STERIS
1D1008 1D1005
STER1D1008EA 0 CNY
STER1D1008 STER1D1005
CIP 100® Detergent System
清洗剂 洗涤剂和表面清洁剂
CIP 100 alkaline detergent is ideal for clean-in-place (CIP) and clean-out-of-place (COP) applications.
  • Low foaming, formulated alkaline cleaner contains biodegradable surfactants
  • Extremely free-rinsing, penetrates, dissolves and removes a broad spectrum of soils using multiple cleaning mechanisms
  • Both specific and non-specific methods are available for detecting cleaning agent residues
High-performance, phosphate-free, alkaline detergent system is for use in manual, immersion, and recirculating spray equipment.
A proprietary blend of potassium hydroxide, surfactants, chelants and other performance-enhancing ingredients provide multiple cleaning mechanisms. The low foaming product removes a wide range of process residues, from fertmentation by-products to silicone-based emulsions and lubricants.
It is specially formulated to meet the unique process cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement and other industries requiring an effective validatable cleaner.
Certifications: Manufactured in a cGMP compliant, FDA registered and ISO 9001/13485 certified facility