Rotary Vane Pumps
供应商: GARDNER DENVER
8912C-01 8917C-80 8925C-02
ILMV8912C-01EA 0 CNY
ILMV8912C-01 ILMV8917C-80 ILMV8925C-02
Rotary Vane Pumps
旋片真空泵
Welch Direct-drive vacuum pumps are portable high performance oil-seal pumps. These pumps come with isolation devices integral to the pump to prevent oil and air contamination of your system in the event of a power failure. Pump can be ordered with an optional exhaust fitler to control exhaust oil mist.
  • Compact for moving from lab to lab
  • Lightweight, compact pump
  • Ultimate to 0,004 mbar (<0,003 torr)
  • Integral isolation valve