DUOSEAL® Vacuum Pumps, Two Stage
供应商: GARDNER DENVER
1397C-03 1397M-01 1400C-01 1400C-02 1402C-01 1402C-02 1402M-01 1405C-02
ILMV1397C-03EA 0 CNY
ILMV1397C-03 ILMV1397M-01 ILMV1400C-01 ILMV1400C-02 ILMV1402C-01 ILMV1402C-02 ILMV1402M-01 ILMV1405C-02
DUOSEAL® Vacuum Pumps, Two Stage
真空泵
The Welch 1397 vacuum pump is a rugged and robust vacuum pump for applications requiring pressures <1 Torr. The 1397 achieves <0,001 Torr with a pumping speed of 500 liters/min (17,7 CFM).
Two stage, oil sealed vacuum pumps should not be operated continuously at pressures above 1 Torr. Operating this vacuum pump at higher pressure will induce excessive oil spraying from the exhaust of the vacuum pump causing premature failure and void warranty.