CHEMSTAR™ Vacuum Pumps
供应商: GARDNER DENVER
1376N-49 1376N-60 1400N-50 1402N-50 1402N-60
ILMV1376N-49EA 0 CNY
ILMV1376N-49 ILMV1376N-60 ILMV1400N-50 ILMV1402N-50 ILMV1402N-60
CHEMSTAR™ Vacuum Pumps
真空泵
CHEMSTAR® pumps are built to withstand corrosive chemical vapor environments - durable performance over the long haul. CHEMSTAR® is designed to minimise the effects of harmful chemicals.
  • CHEMSTAR pump should not be operated continuously at pressures above 10 torr
  • Ultimate pressure measured with a trapped McCleod gauge
  • All single phase motors have overload protection
  • Conduit wiring installation required. No cord, plug or switch provided
Standard filter option possible if the exhaust flange is removed, but not chemically resistant.
Pump rpm: 525
Certifications: Units supplied with CE marking.
Delivery information: Hinged clamp, centering ring assembly and hose adapter are included with all CHEMSTAR pumps.