DUOSEAL® Belt-Drive Vacuum Pumps, Two Stage
供应商: GARDNER DENVER
1374M-01 1376C-03 1376M-01
ILMV1374M-01EA 0 CNY
ILMV1374M-01 ILMV1376C-03 ILMV1376M-01
DUOSEAL® Belt-Drive Vacuum Pumps, Two Stage
真空泵
Vacuum pump is a rugged and robust vacuum pump for applications requiring pressures <1 Torr. Achieves <0,001 Torr.
Two stage, oil sealed vacuum pumps should not be operated continuously at pressures above 1 Torr.
Operating this vacuum pump at higher pressure will induce excessive oil spraying from the exhaust of the vacuum pump causing premature failure and void warranty.