GEM® Direct Drive Vacuum Pumps and Systems
供应商: GARDNER DENVER
8890C-02 8890C-72 8907C-02
ILMV8890C-02EA 0 CNY
ILMV8890C-02 ILMV8890C-72 ILMV8907C-02
GEM® Direct Drive Vacuum Pumps and Systems
真空泵
Direct drive vacuum pumps systems vacuum system is ideal for rotary evaporations with very high boiling point solvents and schlenk lines/vacuum manifolds.
  • Low noise level
  • Anti-suck back valve
  • Forced oil lubrication
The rugged gear pumping mechanism has a wide range of operating pressures and passes vapors readily without the pump mechanism damage often seen in rotary vane pumps. The GEM® efficiently pumps vapors without condensing them into the pump. Provides vacuum to 0,1 torr with free air displacement of 31 L/min.
The complete system comes with a gauge, vacuum regulator for easy vacuum control and an exhaust mist eliminator for recycling oil back into the pump. Ventilation to a hood is recommended.