Premium Vacuum Pump Oils for Direct Drive Vacuum Pumps
供应商: GARDNER DENVER
8995P-15 8995P-20
ILMV8995P-15EA 0 CNY
ILMV8995P-15 ILMV8995P-20
Premium Vacuum Pump Oils for Direct Drive Vacuum Pumps
润滑油
A triple-distilled hydrocarbon oil using severely hydrotreated base stock is designed to resist breakdown at higher RPMs and operating temperatures of direct-drive vacuum pumps. The hydrotreating virtually eliminates aromatics and sulfur to give good resistance to sludge and varnish formation overtime in corrosive environments. Premium oil enables direct-drive vacuum pumps to maintain the highest vacuum performance over time. The oil is light yellow.
  • Low vapor pressure
  • Designated for high RPM direct drive vacuum pumps
  • No additives or inhibitors
  • Recommended for Welch direct-drive vacuum pumps