Standard Duty Dry Vacuum Pump, Wob-L
供应商: GARDNER DENVER
2562C-02 2567C-02
ILMV2562C-02EA 0 CNY
ILMV2562C-02 ILMV2567C-02
Standard Duty Dry Vacuum Pump, Wob-L
真空泵
Welch Standard Duty* vacuum pumps use dry, oil-free WOB-L® Piston technology to deliver industry-leading, strong pumping dynamics that support filtration, drying, aspiration, parts forming, and other rough vacuum applications.
  • Effective for standard duty applications, not strongly acidic, basic or organic vapors
  • Lightweight, compact piston pump
  • Comes with inlet liquid trap, vacuum regulator and muffler
  • Oil-free operation for reduced maintenance
  • High water vapor tolerance
  • Wired for 230 V, 50 Hz, 1 Ph with IEC connection
Welch Standard duty dry vacuum pumps can be used for such vacuum applications as cell culture media sterile filtering/aspiration, DNA/RNA/Plasmid extraction, vacuum ovens, vacuum filtering, vacuum bag parts forming, sampling, cell harvesters, pick and place, package integrity testing, turbidity testing, rice porosity testing, air sampling and many other laboratory and industrial applications.