High Purity Standards Solutions of 68 Element Standard, ICP
供应商: OT CHEMICALS
ICP-MS-68A-A ICP-MS-68A-C ICP-MS-68A-B
XXXXICP-MS-68A-AEA 0 CNY
XXXXICP-MS-68A-A XXXXICP-MS-68A-C XXXXICP-MS-68A-B
High Purity Standards Solutions of 68 Element Standard, ICP
标准品 Alloys and Metal Ore Standards
ICP-MS multi element solution standard at 10 µg/ml with 12 month expiration date.
Solution A of the 3 solution set ICP-MS-68A with 48 components:
Aluminum, arsenic, barium, beryllium, bismuth, boron, cadmium, calcium, cerium, cesium, chromium, cobalt, copper, dysprosium, erbium, europium, gadolinium, gallium, holmium, indium, iron, lanthanum, lead, lithium, lutetium, magnesium, manganese, neodymium, nickel, phosphorus, potassium, praseodymium, rhenium, rubidium, samarium, scandium, selenium, sodium, strontium, terbium, thallium, thorium, thulium, uranium, vanadium, ytterbium, yttrium, zinc at 10 µg/ml in 2% HNO₃
Solution B of the 3 solution set ICP-MS-68A with 13 components:
Antimony, germanium, hafnium, molybdenum, niobium, silicon, silver, tantalum, tellurium, tin, titanium, tungsten, zirconium at 10 µg/ml in 2% HNO₃ + Tr HF
Solution C of the 3 solution set ICP-MS-68A with 7 components:
Gold, iridium, osmium, palladium, platinum, rhodium, ruthenium at 10 µg/ml in 15% HCl
Certifications: This standard is traceable to NIST SRM 3100 series. ISO 9001:2015 certified, ISO/IEC 17025:2017 and ISO 17034:2016 accredited.