间甲基苯硼酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3-甲基苯硼酸

344420010 344420050
ACRO344420010EA 0 CNY
ACRO344420010 ACRO344420050
间甲基苯硼酸 97%
间甲基苯硼酸
分子式: C₇H₉BO₂
分子量: 135.96 g/mol
溶点: 160…162 °C
存储条件: Refrigerator
MDL编号: MFCD00040198
CAS号: 17933-03-8

现在订购

产品技术规格

Appearance White to cream to beige Powder and/or chunks
Infrared spectrum Conforms
Melting point 159 to 165 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR