柠檬醛 cis- and trans-mixture 95%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
3,7-二甲基-2,6-辛二烯醛, 香叶醛和橙花醛混合物
110440050
110441000
110445000
ACRO110440050EA
0
CNY
ACRO110440050
ACRO110441000
ACRO110445000
柠檬醛 cis- and trans-mixture 95%
柠檬醛
分子式:
C₁₀H₁₆O 分子量: 152.24 g/mol 沸点: 229 溶点: –10 °C 密度: 0.889 g/cm³ 闪点: 101 °C 存储条件: Ambient |
MDL编号:
MFCD00006997 CAS号: 5392-40-5 EINECS: 226-394-6 Merck Index: 13,02346 |
产品技术规格
Appearance | Clear yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4860 to 1.4900 (20°C, 589 nm) |
Density | (20/4°C) 0.886 to 0.893 kg/l |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...