3,5-双(三氟甲基)苯硼酸 95%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 3,5-Bis(trifluoromethyl)benzeneboronic acid, (3,5-Bis(trifluoromethyl)phenyl)metaboric acid, (3,5-Bis(trifluoromethyl)phenyl)boronic acid, Dihydroxy(3,5-bis(trifluoromethyl)phenyl)borane

351360010 351360050
ACRO351360010EA 0 CNY
ACRO351360010 ACRO351360050
3,5-双(三氟甲基)苯硼酸 95%
3,5-双(三氟甲基)苯硼酸
分子式: C₈H₅BF₆O₂
分子量: 257.93 g/mol
溶点: 219…226 °C
MDL编号: MFCD00051850
CAS号: 73852-19-4

现在订购

产品技术规格

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 219 to 226 °C
Titration with NaOH 94 to 108 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR