VWR® Crimp Top Headspace Vials, ND20
供应商: VWR Collection
VWRA548-2684EA 780 CNY
VWRA548-2684 VWRA548-2685 VWRA548-2686
VWR® Crimp Top Headspace Vials, ND20
小瓶
Headspace ND20 vials with crimp neck manufactured from 1st hydrolytic class borosilicate glass in both flat bottom and round bottom style. They can be purchased with a variety of aluminium seal styles and septa materials.
  • High performance caps/septa
  • Available with flat DIN crimp neck for tighter seal
  • Thicker wall 1,2 mm withstand higher internal pressure
  • Crimp caps provide a consistently secure seal
  • Beveled top for maximum secure seal
Headspace ND20 vials are available in three different sizes (5, 10 and 20 ml) in amber or clear version. For use in a broad variety of instrument platforms the vials are available with round or a flat bottom.