VWR® Crimp Neck Vials
供应商: VWR Collection
VWRA548-2669EA 3360 CNY
VWRA548-2669 VWRA548-2670
VWR® Crimp Neck Vials
小瓶
Crimp neck ND11 vials are manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of aluminium seal styles and septa materials.
  • Standard vials for GC and HPLC
  • Vials with integrated micro-insert are available
  • Vials have a 40% larger opening than standard opening aluminum seal vials
  • The standard 12×32 mm profile is compatible with 11 mm aluminum seal closures
ND11 crimp neck vials are available in amber or clear version. Multiple crimp cap colours mostly available with assembled stopper or septa are engineered to prevent sample contamination while providing consistent, secure seals. Push-on snap caps for crimp neck vials ND11 are available.