VWR® Snap Ring Vials, ND11, with 11 mm
供应商: VWR Collection
VWRA548-2671EA 3360 CNY
VWRA548-2671 VWRA548-2672
VWR® Snap Ring Vials, ND11, with 11 mm
小瓶
Snap ring ND11 vials manufactured from 1st hydrolytic class borosilicate glass with flat bottom. They can be purchased with a variety of polyethylene seal styles and septa materials.
  • High performance caps/septa
  • Good handling with gloves
  • Quick assembly
  • Wide opening
Vials have a 40% larger opening than standard aluminum seal vials for easier sample filling and to reduce the chance of bent or broken needles during sampling. Vials can also be sealed with an aluminum crimp seal ND11. Available with natural,bule or blue snap ring caps. Available as closed top screw seals or with centre hole
Vials with integrated micro-insert are available.