Anti-Mouse IgG Horse Antibody (HRP)

供应商: CELL SIGNALING TECHNOLOGIES
7076S 7076P2 7076V
CSTA7076SEA 0 CNY
CSTA7076S CSTA7076P2 CSTA7076V
Anti-Mouse IgG Horse Antibody (HRP)
抗体
Type: Secondary
Antigen: Mouse IgG
Clonality:
Clone:
Conjugation: HRP
Epitope:
Host: Horse
Isotype:
Reactivity:
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR