Anti-GAPDH Rabbit Monoclonal Antibody [clone: D16H11]

供应商: CELL SIGNALING TECHNOLOGIES
5174S 5174T
CSTA5174SEA 0 CNY
CSTA5174S CSTA5174T
Anti-GAPDH Rabbit Monoclonal Antibody [clone: D16H11]
抗体
Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: D16H11
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR