Anti-NFkB-p65 (Ser536) phosphate Rabbit Monoclonal Antibody [clone: 93H1]

供应商: CELL SIGNALING TECHNOLOGIES
3033L 3033S 3033T
CSTA3033LEA 0 CNY
CSTA3033L CSTA3033S CSTA3033T
Anti-NFkB-p65 (Ser536) phosphate Rabbit Monoclonal Antibody [clone: 93H1]
抗体
Type: Primary
Antigen: NFkB-p65 (Ser536) phosphate
Clonality: Monoclonal
Clone: 93H1
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Hamster;Monkey;Pig
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR