Anti-IRAK4 Rabbit Polyclonal Antibody

供应商: CELL SIGNALING TECHNOLOGIES
4363S 4363T
CSTA4363SEA 0 CNY
CSTA4363S CSTA4363T
Anti-IRAK4 Rabbit Polyclonal Antibody
抗体
Type: Primary
Antigen: IRAK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Monkey
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR