Anti-Rabbit IgG Goat Antibody (HRP)

供应商: CELL SIGNALING TECHNOLOGIES
7074P2 7074S 7074V
CSTA7074P2EA 0 CNY
CSTA7074P2 CSTA7074S CSTA7074V
Anti-Rabbit IgG Goat Antibody (HRP)
抗体
Type: Secondary
Antigen: Rabbit IgG
Clonality:
Clone:
Conjugation: HRP
Epitope:
Host: Goat
Isotype:
Reactivity: Rabbit
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR