Anti-NFkB-p65 Rabbit Monoclonal Antibody [clone: D14E12]

供应商: CELL SIGNALING TECHNOLOGIES
8242S 8242T
CSTA8242SEA 0 CNY
CSTA8242S CSTA8242T
Anti-NFkB-p65 Rabbit Monoclonal Antibody [clone: D14E12]
抗体
Type: Primary
Antigen: NFkB-p65
Clonality: Monoclonal
Clone: D14E12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Hamster;Monkey;Dog
现在订购

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR