4-(2-氨基乙基)吡啶 ≥97.0% (by titrimetric analysis)
供应商: TCI
|
Danger
|
Synonyms:
2-(4-Pyridyl)ethylamine, 2-Pyridin-4-yl-ethylamine
A1264-25G
A1264-5G
TCIAA1264-25GEA
0
CNY
TCIAA1264-25G
TCIAA1264-5G
4-(2-氨基乙基)吡啶 ≥97.0% (by titrimetric analysis)
4-(2-氨基乙基)吡啶
分子式:
C₇H₁₀N₂ 分子量: 122.17 g/mol 沸点: 120°C 溶点: 76 |
MDL编号:
MFCD00038045 CAS号: 13258-63-4 UN号: 2735 ADR: 8 III |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...