E/E Taxed 99,5 ACS, pure (200 Proof), Riedel-de Haën™
供应商: Honeywell Chemicals
|
Danger
|
Synonyms:
Ethyl alcohol
459844-500ML
459844-2.5L
HONC459844-500MLEA
0
CNY
HONC459844-500ML
HONC459844-2.5L
E/E Taxed 99,5 ACS, pure (200 Proof), Riedel-de Haën™
乙醇
分子式:
C₂H₅OH 分子量: 46.07 g/mol 沸点: 78…79 °C (1.013 hPa) 溶点: –115 °C 密度: 0.790…0.791 g/cm3 (20 °C) 闪点: 12 存储条件: Ambient |
MDL编号:
MFCD00003568 CAS号: 64-17-5 EINECS: 200-578-6 UN号: 1170 ADR: 3 II REACH: 01-2119457610-43 Merck Index: 12,03806 |
产品技术规格
Identity (IR) | complying |
Assay according to ACS (GC) | Min. 99.50 % |
Non-volatile matter | Max. 0.001 % |
Water (Karl Fischer) | Max. 0.2 % |
Titrable Acid | Max. 0.0005 meq/g |
Titrable Base | Max. 0.0002 meq/g |
Solubility in water | complying |
KMnO₄ reducing matter | complying |
Reaction against H₂SO₄ | complying |
Acetone and 2-propanol according to ACS | complying |
Methanol (GC) | Max. 0.1 % |
APHA | Max. 10 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...