甲基丙烯酸, Sigma-Aldrich®

供应商: Merck
Danger

Synonyms: 异丁烯酸, 2-甲基丙烯酸

M-Clarity™ quality level = MQ200

155721-100G 155721-5G
SIAL155721-100GEA 0 CNY
SIAL155721-100G SIAL155721-5G
甲基丙烯酸, Sigma-Aldrich®
甲基丙烯酸
分子式: C₄H₆O₂
沸点: 163 °C (1013 hPa)
溶点: 15 °C
密度: 1,02 g/cm³ (20 °C)
闪点: 68 °C
MDL编号: MFCD00002651
CAS号: 79-41-4
EINECS: 201-204-4
UN号: 2531
ADR: 8,II
Merck Index: 13,05967

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR