N-乙基马来酰亚胺, Sigma-Aldrich®

供应商: Merck
Danger

Synonyms: NEM

M-Clarity™ quality level = MQ200

04259-25G 04259-5G
SIAL04259-25GEA 0 CNY
SIAL04259-25G SIAL04259-5G
N-乙基马来酰亚胺, Sigma-Aldrich®
N-乙基马来酰亚胺
分子式: C₆H₇NO₂
分子量: 125.13 g/mol
沸点: 210
溶点: 46
MDL编号: MFCD00005509
CAS号: 128-53-0
EINECS: 204-892-4
UN号: 2928
ADR: 6.1 II
Merck Index: 13,03855

现在订购


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR