苄硫醇 99%

供应商: Thermo Fisher Scientific
Danger

Synonyms: α-甲苯硫醇, 甲苯硫醇

421261000 421265000
ACRO421261000EA 0 CNY
ACRO421261000 ACRO421265000
苄硫醇 99%
苄硫醇
分子式: C₇H₈S
沸点: 195 °C
溶点: –29 °C
密度: 1,058 g/cm³
闪点: 70 °C
存储条件: Ambient
MDL编号: MFCD00004867
CAS号: 100-53-8
EINECS: 202-862-5
UN号: 2810
ADR: 6.1,II
Merck Index: 12,09456

现在订购

产品技术规格

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5735 to 1.5775 (20°C, 589 nm)
GC ≥98.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR