对甲基苯甲酸甲酯 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
4-Methylbenzoic acid methyl ester, Methyl-4-methylbenzoate, p-Toluic acid methyl ester, 4-Toluic acid methyl ester, Methyl 4-methylbenzoate
414961000
414962500
ACRO414961000EA
0
CNY
ACRO414961000
ACRO414962500
对甲基苯甲酸甲酯 99%
对甲基苯甲酸甲酯
分子式:
C₉H₁₀O₂ 分子量: 150.18 沸点: 220…223 °C 溶点: 32…35 °C 密度: 1.06 g/cm³ 闪点: 90 °C 存储条件: Ambient |
MDL编号:
MFCD00008441 CAS号: 99-75-2 EINECS: 202-784-1 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid or solid |
Infrared spectrum | Conforms |
Melting point | 32 to 35 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...