Isopropyl salicylate 99%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Isopropyl 2-hydroxybenzoate
412891000
412892500
ACRO412891000EA
0
CNY
ACRO412891000
ACRO412892500
Isopropyl salicylate 99%
Isopropyl salicylate
分子式:
C₁₀H₁₂O₃ 沸点: 122 °C (18 mmHg) 密度: 1,06 g/cm³ 闪点: >93 °C 存储条件: Ambient |
MDL编号:
MFCD00035703 CAS号: 607-85-2 EINECS: 210-143-2 |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5090 to 1.5110 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...