N-(3-羟丙基)酞亚胺 99%
供应商: Thermo Fisher Scientific
Synonyms:
2-(3-Hydroxypropyl)isoindoline-1,3-dione
377010250
377011000
ACRO377010250EA
0
CNY
ACRO377010250
ACRO377011000
N-(3-羟丙基)酞亚胺 99%
N-(3-羟丙基)酞亚胺
分子式:
C₁₁H₁₁NO₃ 分子量: 205.21 g/mol 溶点: 75…77 °C |
MDL编号:
MFCD00023097 CAS号: 883-44-3 EINECS: 212-931-1 |
产品技术规格
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 75 to 78 °C |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...