2,4-二甲氧基苄胺 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
1-(2,4-Dimethoxyphenyl)methanamine, (2,4-Dimethoxyphenyl)methanamine
369950050
369950250
ACRO369950050EA
0
CNY
ACRO369950050
ACRO369950250
2,4-二甲氧基苄胺 98%
2,4-二甲氧基苄胺
分子式:
C₉H₁₃NO₂ 沸点: 140 °C (760 mmHg) 密度: 1,113 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00052393 CAS号: 20781-20-8 UN号: 2735 ADR: 8,II |
产品技术规格
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5480 to 1.5500 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...