3-溴苯乙醇 97%
供应商: Thermo Fisher Scientific
|
Warning
|
350560010
350560050
ACRO350560010EA
0
CNY
ACRO350560010
ACRO350560050
3-溴苯乙醇 97%
3-溴苯乙醇
分子式:
C₈H₉BrO 分子量: 201.06 g/mol 沸点: 107…110 °C (1 mmHg) 密度: 1.478 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00191857 CAS号: 28229-69-8 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5730 to 1.5750 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...