5-氟吲哚-2-甲酸 98%

供应商: Thermo Fisher Scientific
Warning

Synonyms: 5-Fluoro-1H-indole-2-carboxylic acid

331590010 331590050
ACRO331590010EA 0 CNY
ACRO331590010 ACRO331590050
5-氟吲哚-2-甲酸 98%
5-氟吲哚-2-甲酸
分子式: C₉H₆FNO₂
分子量: 179.15 g/mol
溶点: 254…258 °C
存储条件: Refrigerator
MDL编号: MFCD00005612
CAS号: 399-76-8
EINECS: 206-919-5

现在订购

产品技术规格

Appearance Yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 254 to 258 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR