乙二胺四乙酸三钾盐二水合物 99+%
供应商: Thermo Fisher Scientific
|
Warning
|
Synonyms:
乙二胺四乙酸三钾, 依地酸, EDTA
301601000
301605000
ACRO301601000EA
0
CNY
ACRO301601000
ACRO301605000
乙二胺四乙酸三钾盐二水合物 99+%
乙二胺四乙酸三钾盐二水合物
分子式:
C₁₀H₁₃K₃N₂O₈·2H₂O 分子量: 442.55 g/mol 存储条件: Ambient |
MDL编号:
MFCD00149672 CAS号: 65501-24-8 EINECS: 241-543-5 |
产品技术规格
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥99.0 % (on dry substance) |
Water | ≤9 % (K.F.) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...