2-氯苯基乙醇 98%
供应商: Thermo Fisher Scientific
186190050
186190250
ACRO186190050EA
0
CNY
ACRO186190050
ACRO186190250
2-氯苯基乙醇 98%
2-氯苯基乙醇
分子式:
C₈H₉ClO 沸点: 84…85 °C (3 mmHg) 密度: 1,19 g/cm³ 闪点: >110 °C 存储条件: Ambient |
MDL编号:
MFCD00002888 CAS号: 19819-95-5 EINECS: 243-348-0 |
产品技术规格
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5495 to 1.5515 (20°C, 589 nm) |
GC | ≥97.5 % |
Residue after ignition | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...