氯甲酸丁酯 98%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Chloroformic acid butyl ester, n-Butyl chloroformate
180170010
180170250
180171000
ACRO180170010EA
0
CNY
ACRO180170010
ACRO180170250
ACRO180171000
氯甲酸丁酯 98%
氯甲酸丁酯
分子式:
C₅H₉ClO₂ 沸点: 142 °C (1013 hPa) 溶点: –70 °C 密度: 1,06 g/cm³ 闪点: 39 °C 存储条件: Refrigerator |
MDL编号:
MFCD00000650 CAS号: 592-34-7 EINECS: 209-750-5 UN号: 2743 ADR: 6.1,II |
产品技术规格
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4110 to 1.4130 (20°C, 589 nm) |
GC | ≥97.5 % |
Color scale | ≤100 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...