1,1,4,4-四苯基-1,3-丁二烯 99%

供应商: Thermo Fisher Scientific
Warning

Synonyms: TPB

176540050 176540010
ACRO176540050EA 0 CNY
ACRO176540050 ACRO176540010
1,1,4,4-四苯基-1,3-丁二烯 99%
1,1,4,4-四苯基-1,3-丁二烯
分子式: (C₆H₅)₂CCHCHC(C₆H₅)₂
分子量: 358.48 g/mol
沸点: 556
溶点: 196…200 °C
存储条件: Ambient
MDL编号: MFCD00004766
CAS号: 1450-63-1
EINECS: 215-914-7

现在订购

产品技术规格

Appearance White to light yellow Powder or needles
Infrared spectrum Conforms
Melting point 195 to 200 °C
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR