4-(溴甲基)苯甲酸 97%

供应商: Thermo Fisher Scientific
Warning

Synonyms: α-溴对甲苯酸

170400100 170400500
ACRO170400100EA 0 CNY
ACRO170400100 ACRO170400500
4-(溴甲基)苯甲酸 97%
4-(溴甲基)苯甲酸
分子式: C₈H₇BrO₂
分子量: 215.05 g/mol
沸点: 316 °C (1013 hPa)
溶点: 223…227 °C
存储条件: Ambient
MDL编号: MFCD00002567
CAS号: 6232-88-8
EINECS: 228-343-3

现在订购

产品技术规格

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR