3-硝基邻苯二甲酸酐 97%
供应商: Thermo Fisher Scientific
|
Warning
|
168990250
168991000
ACRO168990250EA
0
CNY
ACRO168990250
ACRO168991000
3-硝基邻苯二甲酸酐 97%
3-硝基邻苯二甲酸酐
分子式:
C₈H₃NO₅ 溶点: 160…165 °C |
MDL编号:
MFCD00005921 CAS号: 641-70-3 EINECS: 211-373-6 |
产品技术规格
Appearance | Yellow to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 160 to 165 °C |
Titration by coupling | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...