噻唑 99%

供应商: Thermo Fisher Scientific
Warning

166130010 166130050
ACRO166130010EA 0 CNY
ACRO166130010 ACRO166130050
噻唑 99%
噻唑
分子式: C₃H₃NS
分子量: 85.13 g/mol
沸点: 117…118 °C (760 mmHg)
溶点: -34
密度: 1.198 g/cm³
闪点: 26 °C
存储条件: Ambient
MDL编号: MFCD00005315
CAS号: 288-47-1
EINECS: 206-021-3
UN号: 1993
ADR: 3 III
Merck Index: 12,09442

现在订购

产品技术规格

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5365 to 1.5395 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR