4-(溴甲基)苯甲腈 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 4-氰基苄溴, α-溴对甲基苯甲腈

162760100 162760500
ACRO162760100EA 0 CNY
ACRO162760100 ACRO162760500
4-(溴甲基)苯甲腈 98%
4-(溴甲基)苯甲腈
分子式: C₈H₆BrN
分子量: 196.05 g/mol
沸点: 135…137 °C (0.6 mmHg)
溶点: 113…117 °C
MDL编号: MFCD00001829
CAS号: 17201-43-3
EINECS: 241-246-0
UN号: 1759
ADR: 8 II

现在订购

产品技术规格

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 113 to 118 °C
GC ≥97.5 %
Iron (Fe) ≤0.1 %
Phosphorus (P) ≤0.04 %
Potassium (K) ≤0.4 %
Silicon (Si) ≤0.06 %
Sodium (Na) ≤0.3 %
Sulfur (S) ≤0.07 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR