巴豆酸甲酯 96%

供应商: Thermo Fisher Scientific
Danger

Synonyms: Methyl trans-2-butenoate, 2-Butenoic acid methyl ester, trans-Methyl crotonate, trans-Crotonic acid methyl ester, Crotonic acid methyl ester, trans-2-Butenoic acid methyl ester

160840050 160841000 160845000
ACRO160840050EA 0 CNY
ACRO160840050 ACRO160841000 ACRO160845000
巴豆酸甲酯 96%
巴豆酸甲酯
分子式: C₅H₈O₂
分子量: 100.12 g/mol
沸点: 118
密度: 0.94 g/cm³
闪点: –1 °C
MDL编号: MFCD00009287
CAS号: 623-43-8
EINECS: 210-793-7
UN号: 3272
ADR: 3 II
Merck Index: 13,02625

现在订购

产品技术规格

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4230 to 1.4270 (20°C, 589 nm)
GC ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR