马来酰亚胺 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: 2,5-吡咯二酮

156710050 156710100 156710250
ACRO156710050EA 0 CNY
ACRO156710050 ACRO156710100 ACRO156710250
马来酰亚胺 98%
马来酰亚胺
分子式: C₄H₃NO₂
沸点: 102 - 105 °C (55 -rr)
溶点: 90…94 °C
密度: 1,2493 g/cm³ (106 °C)
存储条件: Ambient
MDL编号: MFCD00005494
CAS号: 541-59-3
EINECS: 208-787-4
UN号: 2923
ADR: 8,III

现在订购

产品技术规格

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 90 to 94 °C
GC ≥97.5 %
Titration with NaOH ≥97.5 %
Substance darkened by H2SO4 Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR