N,N-二乙基三甲基甲硅烷胺 98+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
N-(三甲基甲硅烷基)二乙胺, (二乙氨基)三甲基硅烷, TMSDEA
155690250
155691000
ACRO155690250EA
0
CNY
ACRO155690250
ACRO155691000
N,N-二乙基三甲基甲硅烷胺 98+%
N,N-二乙基三甲基甲硅烷胺
分子式:
C₇H₁₉NSi 分子量: 145.32 g/mol 沸点: 125…126 °C (760 mmHg) 溶点: –10 °C 密度: 0.76 g/cm³ 闪点: 10 °C 存储条件: Ambient |
MDL编号:
MFCD00009040 CAS号: 996-50-9 EINECS: 213-637-6 UN号: 2733 ADR: 3 II |
产品技术规格
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4103 to 1.4123 (20°C, 589 nm) |
GC | ≥98.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...