(3,4-二甲氧苯基)乙腈 99+%
供应商: Thermo Fisher Scientific
|
Danger
|
Synonyms:
3,4-二甲氧基苯乙腈, 高藜芦腈, (3,4-二甲氧苯基)乙腈
155260250
155261000
ACRO155260250EA
0
CNY
ACRO155260250
ACRO155261000
(3,4-二甲氧苯基)乙腈 99+%
(3,4-二甲氧苯基)乙腈
分子式:
C₁₀H₁₁NO₂ 分子量: 177.2 沸点: 171…178 °C (10 mmHg) 溶点: 62…65 °C 闪点: 250 °C 存储条件: Ambient |
MDL编号:
MFCD00001911 CAS号: 93-17-4 EINECS: 202-225-1 UN号: 2811 ADR: 6.1 III |
产品技术规格
Appearance | White to off-white Crystals or crystalline needles |
Infrared spectrum | Conforms |
Melting point | 62 to 65 °C |
Loss on drying | ≤0.5 % (vacuum) |
Sulfated ash | ≤0.1 % |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...