N-丁基二乙醇胺 98%

供应商: Thermo Fisher Scientific
Danger

Synonyms: N,N-双(2-羟乙基)丁胺, 2,2′-丁基亚氨基二乙醇

154360010 154362500
ACRO154360010EA 0 CNY
ACRO154360010 ACRO154362500
N-丁基二乙醇胺 98%
N-丁基二乙醇胺
分子式: H₃CCH₂CH₂CH₂N(CH₂CH₂OH)₂
沸点: 264…278 °C (760 mmHg)
溶点: –70 °C
密度: 0,968 g/cm³
闪点: 143 °C
存储条件: Ambient
MDL编号: MFCD00002856
CAS号: 102-79-4
EINECS: 203-055-0
UN号: 2735
ADR: 8,II

现在订购

产品技术规格

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4610 to 1.4640 (20°C, 589 nm)
Water ≤0.5 %
GC ≥97.5 %
Color scale ≤200 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR